siRNA Details
Gene Symbol: Trpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039104.2 | NP_001034193.2 | 4 | 697 | ||
NM_018752.3 | NP_061222.3 | 4 | 590 | |||
GenBank mRNA | AF047714.1 | - | 4 | 459 | ||
AJ867481.1 | - | 4 | 604 | |||
AK044507.1 | - | 4 | 688 | |||
AK148094.1 | - | 4 | 495 | |||
AK148193.1 | - | 4 | 498 | |||
AK148268.1 | - | 4 | 444 | |||
AK148275.1 | - | 4 | 500 | |||
AK149290.1 | - | 4 | 619 | |||
AK164395.1 | - | 4 | 500 | |||
AK164423.1 | - | 4 | 507 | |||
AK184260.1 | - | 2 | 256 | |||
AK185201.1 | - | 3 | 252 | |||
AK190970.1 | - | 2 | 246 | |||
AK191452.1 | - | 2 | 246 | |||
AK191953.1 | - | 2 | 246 | |||
AK193027.1 | - | 2 | 246 | |||
AK193309.1 | - | 2 | 246 | |||
AK193599.1 | - | 2 | 247 | |||
AK193876.1 | - | 2 | 246 | |||
AK194310.1 | - | 2 | 246 | |||
AK195176.1 | - | 2 | 246 | |||
AK195666.1 | - | 2 | 246 | |||
AK198002.1 | - | 2 | 246 | |||
AK198830.1 | - | 336 | ||||
AK199687.1 | - | 2 | 247 | |||
AK199999.1 | - | 2 | 246 | |||
AK200915.1 | - | 2 | 246 | |||
AK201913.1 | - | 2 | 246 | |||
AK202423.1 | - | 2 | 246 | |||
AK203591.1 | - | 2 | 248 | |||
AK203858.1 | - | 2 | 246 | |||
AK203906.1 | - | 1 | 45 | |||
AK204991.1 | - | 2 | 198 | |||
AK205201.1 | - | 1 | 148 | |||
AK206340.1 | - | 3 | 252 | |||
AK212724.1 | - | 1 | 117 | |||
AY180104.2 | - | 4 | 697 | |||
BC082560.1 | - | 4 | 466 | |||
BC085168.1 | - | 3 | 269 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
