siRNA Details
Gene Symbol: Phc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195130.1 | NP_001182059.1 | 4 | 544 | ||
NM_018774.4 | NP_061244.1 | 4 | 537 | |||
GenBank mRNA | BC057571.1 | - | 4 | 480 | 6410302 | |
U81491.1 | - | 3 | 434 |
