siRNA Details
Gene Symbol: Tbc1d8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018775.4 | NP_061245.3 | 6 | 1188 | ||
GenBank mRNA | AK138630.1 | - | 6 | 1186 | ||
AK141520.1 | - | 6 | 1177 | |||
AK141816.1 | - | 6 | 1142 | |||
BC005421.1 | - | 3 | 523 | |||
BC065081.1 | - | 6 | 1122 | 6832495 | ||
U88873.1 | - | 2 | 408 |
