siRNA Details
Gene Symbol: Hs3st3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018805.2 | NP_061275.2 | 2 | 984 | ||
GenBank mRNA | AF168992.1 | - | 2 | 942 | ||
AK085242.1 | - | 2 | 993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
