siRNA Details
Gene Symbol: Hs3st3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018805.2 | NP_061275.2 | 2 | 1062 | ||
GenBank mRNA | AF168992.1 | - | 2 | 1020 | ||
AK085242.1 | - | 2 | 1071 | |||
AK171996.1 | - | 2 | 1056 | |||
AK172015.1 | - | 2 | 1061 | |||
AK172271.1 | - | 2 | 1063 | |||
BC116733.1 | - | 2 | 929 | 40126052 | ||
BC116743.1 | - | 2 | 929 | 40126062 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191646.
Gene Ontology Categories:
