siRNA Details
Gene Symbol: Cyhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276321.1 | NP_001263250.1 | 2 | 762 | ||
NM_019396.3 | NP_062269.1 | 3 | 695 | |||
GenBank mRNA | AJ251516.1 | - | 3 | 548 | ||
AK080867.1 | - | 2 | 783 | |||
AK129156.1 | - | 1 | 1869 | |||
BC039646.1 | - | 1 | 88 | |||
BC062184.1 | - | 2 | 751 | 6515695 | ||
BC082772.1 | - | 2 | 762 | 6508580 | ||
BE653118.1 | - | 1 | 121 | |||
BX512375.1 | - | 2 | 301 | |||
BY287678.1 | - | 2 | 326 | |||
CF742880.1 | - | 1 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
