siRNA Details
Gene Symbol: Ankef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175667.4 | NP_783598.1 | 1 | 23 | ||
GenBank mRNA | AK029804.1 | - | 159 | |||
BC089504.1 | - | 1 | 64 | |||
BC120825.1 | - | 1 | 23 | 40129748 | ||
BC144833.1 | - | 1 | 23 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
