siRNA Details
Gene Symbol: Irgm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019440.3 | NP_062313.3 | 2 | 822 | ||
GenBank mRNA | AJ007972.1 | - | 1 | 623 | ||
AK142291.1 | - | 2 | 824 | |||
AK152870.1 | - | 2 | 763 | |||
AK153309.1 | - | 2 | 763 | |||
AK165347.1 | - | 2 | 760 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
