siRNA Details
Gene Symbol: Lgals8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199043.1 | NP_001185972.1 | 10 | 2745 | ||
NM_001291055.1 | NP_001277984.1 | 9 | 2274 | |||
NM_001291057.1 | NP_001277986.1 | 9 | 2470 | |||
NM_001291060.1 | NP_001277989.1 | 9 | 2416 | |||
NM_018886.5 | NP_061374.1 | 10 | 2559 | |||
GenBank mRNA | AK004782.2 | - | 10 | 2427 | ||
AK075736.1 | - | 9 | 2226 | |||
AK083350.1 | - | 10 | 2312 | |||
AK155066.1 | - | 9 | 2185 | |||
BC040243.1 | - | 10 | 2393 | 2582159 |
Gene Symbol: Gpr85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145066.4 | NP_659503.1 | 2 | 761 | ||
GenBank mRNA | BC065154.1 | - | 2 | 761 | 6418997 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
