siRNA Details
Gene Symbol: Cyhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019396.3 | NP_062269.1 | 5 | 1434 | ||
GenBank mRNA | AJ251516.1 | - | 5 | 1287 | ||
AK028994.1 | - | 1 | 193 | |||
BC039646.1 | - | 3 | 827 | |||
CF740409.1 | - | 1 | 224 | |||
CN527531.1 | - | 2 | 579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
