siRNA Details
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284341.1 | NP_001271270.1 | 10 | 1843 | ||
NM_001284343.1 | NP_001271272.1 | 6 | 921 | |||
NM_007270.4 | NP_009201.2 | 9 | 1684 | |||
GenBank mRNA | AB209352.1 | - | 3 | 1068 | ||
AF089745.1 | - | 9 | 1355 | |||
AK075331.1 | - | 9 | 1648 | |||
AK128597.1 | - | 10 | 1625 | |||
AK293440.1 | - | 6 | 899 | |||
AK293954.1 | - | 8 | 1257 | |||
AK294266.1 | - | 8 | 1468 | |||
AK300328.1 | - | 10 | 1806 | |||
AK304643.1 | - | 6 | 759 | |||
AK316290.1 | - | 4 | 574 | |||
AL050187.1 | - | 1 | 14 | |||
BC007443.2 | - | 3 | 400 | |||
BC064418.1 | - | 4 | 607 | |||
BC072422.1 | - | 3 | 351 | |||
BC099918.1 | - | 2 | 246 | |||
BC101723.1 | - | 9 | 1612 | 8069229 | ||
BC112053.1 | - | 9 | 1612 | 8327521 |
Gene Symbol: FKBP9P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003949.1 | 5 | 663 | |||
NR_027339.1 | 3 | 767 | ||||
NR_027340.1 | 2 | 396 | ||||
NR_027342.1 | 3 | 677 | ||||
GenBank mRNA | AK289438.1 | - | 2 | 352 | ||
AK312981.1 | - | 2 | 396 | |||
BC011872.2 | - | 3 | 677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
