siRNA Details
Gene Symbol: Tro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002272.3 | NP_001002272.1 | 12 | 3936 | ||
NM_001290770.1 | NP_001277699.1 | 12 | 3885 | |||
GenBank mRNA | AF241244.1 | - | 13 | 3740 | ||
AF319977.1 | - | 12 | 3759 | |||
AK122449.1 | - | 12 | 2766 | |||
BC053018.1 | - | 1 | 1486 | |||
BC075630.1 | - | 12 | 3885 | 30536880 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
