siRNA Details
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019492.3 | NP_062365.2 | 5, 6 | 456 | ||
GenBank mRNA | AJ250999.1 | - | 5, 6 | 456 | ||
AK015722.1 | - | 3, 4 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
