siRNA Details
Gene Symbol: Car5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181315.4 | NP_851832.2 | 8 | 944 | ||
GenBank mRNA | AF192978.1 | - | 8 | 965 | ||
AK085670.1 | - | 8 | 919 | |||
BC034413.1 | - | 8 | 895 | 4221296 |

More Information
Other Species Targeted

Gene Ontology Categories:
