siRNA Details
Gene Symbol: Tro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002272.3 | NP_001002272.1 | 12 | 5859 | ||
NM_001290770.1 | NP_001277699.1 | 12 | 5808 | |||
GenBank mRNA | AF241244.1 | - | 14 | 5423 | ||
AF319977.1 | - | 13 | 5442 | |||
AK032791.1 | - | 1 | 1548 | |||
AK122449.1 | - | 12 | 4689 | |||
BC053018.1 | - | 1 | 3409 | |||
BC075630.1 | - | 12 | 5808 | 30536880 |

More Information
Other Species Targeted

Gene Ontology Categories:
