siRNA Details
Gene Symbol: Plxnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019587.2 | NP_062533.2 | 34, 35 | 5652 | ||
GenBank mRNA | AJ627037.1 | - | 6, 7 | 681 | ||
AK122467.1 | - | 28, 29 | 4880 | |||
AK138168.1 | - | 30, 31 | 4657 | |||
AK138631.1 | - | 28, 29 | 4833 | |||
AK140240.1 | - | 32, 33 | 5834 | |||
AK170004.1 | - | 21, 22 | 2903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
