siRNA Details
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019694.1 | NP_062668.1 | 3 | 505 | ||
GenBank mRNA | AB041582.1 | - | 3 | 518 | ||
AF061026.1 | - | 3 | 505 | |||
AK133849.1 | - | 3 | 524 | |||
BC023862.1 | - | 3 | 502 | |||
BC046326.1 | - | 3 | 512 | |||
BC055865.1 | - | 3 | 523 | |||
BC061115.1 | - | 3 | 518 | 6533034 | ||
BC061690.1 | - | 3 | 505 |
