siRNA Details
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF154120.1 | - | 3, 4 | 526 | ||
AK154079.1 | - | 3, 4 | 435 | |||
AK167993.1 | - | 3, 4 | 417 | |||
AK219007.1 | - | 1, 2 | 52 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
