siRNA Details
Gene Symbol: Adar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF052506.1 | - | 2 | 529 | ||
AF291050.1 | - | 2 | 523 | |||
AF291876.1 | - | 2 | 523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
