siRNA Details
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019666.2 | NP_062640.2 | 11 | 2469 | ||
GenBank mRNA | AF093821.1 | - | 10 | 2127 | ||
AF408434.1 | - | 9 | 1929 | |||
BC055863.1 | - | 11 | 2247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
