siRNA Details
Gene Symbol: Ppp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019674.3 | NP_062648.1 | 6 | 488 | ||
GenBank mRNA | AF088911.1 | - | 6 | 513 | ||
AK027923.1 | - | 4 | 267 | |||
AK144244.1 | - | 5 | 415 | |||
U79747.1 | - | 2 | 140 |

More Information
Other Species Targeted

Gene Ontology Categories:
