siRNA Details
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028920.1 | - | 14 | 2235 | ||
AK004801.1 | - | 14 | 2241 | |||
AK077864.1 | - | 1 | 225 | |||
AK089092.1 | - | 14 | 2248 | |||
BC042602.1 | - | 14 | 3118 | |||
BC054433.1 | - | 12 | 2815 | |||
BC060170.1 | - | 14 | 3115 | |||
BC060209.1 | - | 13 | 2731 | |||
BC078448.1 | - | 13 | 2621 | |||
BC125521.1 | - | 14 | 2231 | 40130036 | ||
BC137746.1 | - | 14 | 2231 | 8860765 |

More Information
Other Species Targeted

