siRNA Details
Gene Symbol: PIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006221.3 | NP_006212.1 | 4 | 991 | ||
NR_038422.2 | 5 | 1044 | ||||
NR_038830.1 | 6 | 713 | ||||
XM_011528068.2 | XP_011526370.1 | 6 | 982 | |||
GenBank mRNA | AF086512.1 | - | 4 | 389 | ||
AK092970.1 | - | 3 | 3003 | |||
AK097558.1 | - | 4 | 1527 | |||
AK291074.1 | - | 4 | 875 | |||
BC002899.1 | - | 4 | 880 | 3941595 | ||
BC031971.1 | - | 6 | 1551 | |||
BG912418.1 | - | 6 | 583 | |||
U49070.1 | - | 4 | 877 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
