siRNA Details
Gene Symbol: Cript | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019936.3 | NP_064320.1 | 3 | 202 | ||
GenBank mRNA | AF151637.1 | - | 3 | 167 | ||
AK004828.1 | - | 3 | 195 | |||
AK008523.1 | - | 3 | 187 | |||
AK029409.1 | - | 3 | 190 | |||
AK213321.1 | - | 1 | 34 | |||
BC002258.1 | - | 2 | 84 | |||
BC023013.1 | - | 3 | 176 | 5369938 | ||
BY123761.1 | - | 3 | 203 |
