siRNA Details
Gene Symbol: Ccnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019937.3 | NP_064321.2 | 3, 4 | 604 | ||
GenBank mRNA | AF159159.1 | - | 3, 4 | 557 | ||
AF467251.1 | - | 3, 4 | 598 | |||
AK011629.1 | - | 3, 4 | 561 | |||
AK051380.1 | - | 3, 4 | 561 | |||
AK194826.1 | - | 1, 2 | 21 | |||
BC037204.1 | - | 3, 4 | 542 | |||
BC094383.1 | - | 3, 4 | 540 | 30604933 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053662, AF030091, BC161817.
Gene Ontology Categories:
