siRNA Details
Gene Symbol: Exoc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198103.2 | NP_932771.1 | 1 | 999 | ||
GenBank mRNA | BC057052.1 | - | 1 | 999 | 6822067 |
Gene Symbol: Mnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019944.2 | NP_064328.2 | 3 | 1277 | ||
GenBank mRNA | AF153046.1 | - | 1277 | |||
BC132283.1 | - | 3 | 1190 | 40130560 | ||
BC144718.1 | - | 3 | 1191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
