siRNA Details
Gene Symbol: Pnkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039509.1 | NP_001034598.1 | 3 | 364 | ||
NM_019999.2 | NP_064383.1 | 2 | 273 | |||
GenBank mRNA | AB041609.1 | - | 2 | 274 | ||
AK041238.1 | - | 2 | 207 | |||
AK173125.1 | - | 3 | 338 | |||
BC058945.1 | - | 4 | 460 | 6812127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
