siRNA Details
Gene Symbol: Dcpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC024627.1 | - | 261 | 4925312 | ||
BC045522.1 | - | 4 | 271 | 4190591 | ||
BC090399.1 | - | 261 | 4168483 | |||
DQ237936.1 | - | 279 | ||||
S76879.1 | - | 320 | ||||
U03711.1 | - | 284 |
