siRNA Details
Gene Symbol: Mnx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019944.2 | NP_064328.2 | 3 | 1283 | ||
GenBank mRNA | AF153046.1 | - | 1283 | |||
BC132283.1 | - | 3 | 1196 | 40130560 | ||
BC144718.1 | - | 3 | 1197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
