siRNA Details
Gene Symbol: Vmn2r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104592.1 | NP_001098062.1 | 5 | 2184 |
Gene Symbol: Vmn2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019918.2 | NP_064302.2 | 6 | 2439 | ||
GenBank mRNA | AB267725.1 | - | 6 | 2439 | ||
AF053986.1 | - | 6 | 2439 |
Gene Symbol: Vmn2r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104614.1 | NP_001098084.1 | 6 | 2370 |
Gene Symbol: Vmn2r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104615.1 | NP_001098085.1 | 5 | 2184 |
Gene Symbol: Vmn2r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104618.1 | NP_001098088.1 | 5 | 2184 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
