siRNA Details
Gene Symbol: Krt71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019956.1 | NP_064340.1 | 1 | 467 | ||
GenBank mRNA | AB033744.1 | - | 1 | 467 | ||
AB100413.1 | - | 1 | 467 | |||
AK003610.1 | - | 1 | 479 | |||
AK003704.1 | - | 1 | 479 | |||
AY033497.1 | - | 1 | 467 | |||
BC125346.1 | - | 1 | 462 | 40129861 | ||
BC125348.1 | - | 1 | 462 | 40129863 |
Gene Symbol: Krt72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213728.1 | NP_998893.1 | 1 | 425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
