siRNA Details
Gene Symbol: Zfp109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020262.3 | NP_064658.1 | 3, 4 | 712 | ||
GenBank mRNA | AF167316.1 | - | 3, 4 | 674 | ||
BC119619.1 | - | 2, 3 | 281 | 40062160 | ||
BC119620.1 | - | 3, 4 | 339 | 40062167 |
