siRNA Details
Gene Symbol: Calm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020036.4 | NP_064420.2 | 1 | 297 | ||
GenBank mRNA | AK003648.1 | - | 1 | 298 | ||
AK009664.1 | - | 1 | 299 | |||
AK009956.1 | - | 1 | 298 | |||
AY293058.1 | - | 1 | 280 | |||
BC060284.1 | - | 1 | 283 | 30293362 | ||
BC145379.1 | - | 1 | 253 | 9053937 |
Gene Symbol: Calm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008706.2 | NP_001008706.1 | 2 | 214 | ||
GenBank mRNA | AY314008.1 | - | 119 | |||
BC138691.1 | - | 1 | 253 | 8861713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
