siRNA Details
Gene Symbol: Tssc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115085.1 | NP_001108557.1 | 3 | 736 | ||
NM_020285.2 | NP_064681.1 | 3 | 766 | |||
GenBank mRNA | AB038011.1 | - | 3 | 766 | ||
AB041597.1 | - | 1 | 1107 | |||
AJ279796.1 | - | 3 | 766 | |||
AK088052.1 | - | 3 | 701 | |||
CX239537.1 | - | 3 | 680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
