siRNA Details
Gene Symbol: Glra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290821.1 | NP_001277750.1 | 2, 3 | 657 | ||
NM_020492.4 | NP_065238.2 | 2, 3 | 657 | |||
GenBank mRNA | AK078258.1 | - | 2, 3 | 657 | ||
AY129229.1 | - | 2, 3 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
