siRNA Details
Gene Symbol: ASIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308804.1 | - | 5 | 1671 | ||
AK309373.1 | - | 5 | 1666 | |||
BC031812.1 | - | 3 | 2405 | 4944238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
