siRNA Details
Gene Symbol: Clec2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289706.1 | NP_001276635.1 | 7 | 1056 | ||
NM_001289707.1 | NP_001276636.1 | 6 | 981 | |||
NM_001289708.1 | NP_001276637.1 | 5 | 898 | |||
NM_020257.2 | NP_064653.1 | 6 | 969 | |||
GenBank mRNA | AF121352.1 | - | 6 | 942 | ||
AK041724.1 | - | 5 | 1684 | |||
AY137344.1 | - | 6 | 980 | |||
AY137345.1 | - | 6 | 968 | |||
AY256574.1 | - | 7 | 1047 | |||
AY256575.1 | - | 6 | 970 | |||
AY256576.1 | - | 5 | 887 |
