siRNA Details
Gene Symbol: Tssc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115085.1 | NP_001108557.1 | 3 | 915 | ||
NM_020285.2 | NP_064681.1 | 3 | 945 | |||
GenBank mRNA | AB038011.1 | - | 3 | 945 | ||
AB041597.1 | - | 1 | 1286 | |||
AJ279796.1 | - | 3 | 945 | |||
AK088052.1 | - | 3 | 879 |

More Information
Other Species Targeted

Gene Ontology Categories:
