siRNA Details
Gene Symbol: Olfr747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207156.2 | NP_997039.2 | 1 | 903 |
Gene Symbol: Olfr749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020288.2 | NP_064684.2 | 2 | 969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
