siRNA Details
Gene Symbol: Tbx20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020496.3 | NP_065242.1 | 7 | 1361 | ||
GenBank mRNA | AF260557.1 | - | 7 | 1366 | ||
AJ277486.1 | - | 7 | 903 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
