siRNA Details
Gene Symbol: Retnlg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181596.4 | NP_853627.2 | 2 | 228 | ||
GenBank mRNA | AB086819.1 | - | 1 | 149 | ||
AJ514933.1 | - | 1 | 152 | |||
AJ536019.1 | - | 2 | 191 | |||
AK158401.1 | - | 2 | 192 | |||
BC117106.1 | - | 1 | 141 | 40126425 | ||
DY246276.1 | - | 2 | 260 |
Gene Symbol: Retnla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020509.3 | NP_065255.2 | 2 | 212 | ||
GenBank mRNA | AF205951.1 | - | 1 | 145 | ||
AF290872.1 | - | 2 | 250 | |||
AF316397.2 | - | 2 | 154 | |||
AF323082.1 | - | 2 | 222 | |||
AK007553.1 | - | 2 | 157 | |||
BC029248.1 | - | 1 | 152 | 4189285 |

More Information
Other Species Targeted

Gene Ontology Categories:
