siRNA Details
Gene Symbol: Olfr1507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170918.1 | NP_001164389.1 | 2 | 1011 | ||
NM_020512.2 | NP_065258.2 | 1 | 518 | |||
GenBank mRNA | BC119163.1 | - | 1 | 518 | 8733912 | |
BC119165.1 | - | 1 | 518 | 8733914 |
