siRNA Details
Gene Symbol: Ptprz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133917.1 | - | 2103 | |||
AK155419.1 | - | 435 | ||||
AK156638.1 | - | 480 |
Gene Symbol: Cdk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155129.1 | - | 187 |

More Information
Other Species Targeted

