siRNA Details
Gene Symbol: Retnla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020509.3 | NP_065255.2 | 2 | 218 | ||
GenBank mRNA | AF205951.1 | - | 1 | 151 | ||
AF290872.1 | - | 2 | 256 | |||
AF316397.2 | - | 2 | 160 | |||
AF323082.1 | - | 2 | 228 | |||
AK007553.1 | - | 2 | 163 | |||
BC029248.1 | - | 1 | 158 | 4189285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
