siRNA Details
Gene Symbol: Lenep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020517.4 | NP_065263.1 | 2 | 453 | ||
GenBank mRNA | AK053703.1 | - | 466 | |||
BC006806.1 | - | 683 | 3152957 | |||
CK627663.1 | - | 2 | 416 |
Gene Symbol: Flad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK044501.1 | - | 1902 | |||
BC009152.1 | - | 1475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
