siRNA Details
Gene Symbol: Terf2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020584.2 | NP_065609.2 | 3 | 2452 | ||
GenBank mRNA | AB041557.1 | - | 4 | 1342 | ||
AB041559.1 | - | 4 | 1343 | |||
AK148508.1 | - | 3 | 2412 | |||
AK148537.1 | - | 3 | 2414 | |||
AK165074.1 | - | 3 | 2410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
