siRNA Details
Gene Symbol: Tk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021028.3 | NP_066356.3 | 6 | 449 | ||
GenBank mRNA | AF105217.1 | - | 6 | 424 | ||
AJ249341.2 | - | 6 | 427 | |||
AK041651.1 | - | 6 | 441 | |||
AK087557.1 | - | 6 | 450 | |||
AK154160.1 | - | 6 | 443 | |||
AK187899.1 | - | 2 | 78 | |||
AK196494.1 | - | 2 | 74 | |||
AK202106.1 | - | 2 | 74 | |||
AK216095.1 | - | 2 | 74 |

More Information
Other Species Targeted

Gene Ontology Categories:
