siRNA Details
Gene Symbol: Rhox4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039698.1 | NP_001034787.1 | 2 | 533 | ||
GenBank mRNA | AJ972671.1 | - | 2 | 533 |
Gene Symbol: Rhox4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021300.2 | NP_067275.1 | 2 | 441 | ||
GenBank mRNA | AF265350.1 | - | 2 | 429 | ||
AJ972666.1 | - | 2 | 441 | |||
BC172126.1 | - | 2 | 388 | 9054796 |
Gene Symbol: Rhox4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039695.1 | NP_001034784.1 | 314 | |||
GenBank mRNA | AB221641.1 | - | 310 | |||
AJ972668.1 | - | 314 | ||||
BC147696.1 | - | 2 | 388 | 9056669 |
Gene Symbol: Rhox4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201236.3 | NP_957688.2 | 2 | 532 | ||
GenBank mRNA | AJ972669.1 | - | 2 | 532 | ||
BC094891.1 | - | 2 | 494 | 30019143 | ||
BC172128.1 | - | 388 | 9054798 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
