siRNA Details
Gene Symbol: Cd200r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021325.3 | NP_067300.1 | 3 | 443 | ||
GenBank mRNA | AK089173.1 | - | 3 | 380 | ||
AK089415.1 | - | 4 | 674 | |||
AK154325.1 | - | 3 | 434 | |||
AK154722.1 | - | 3 | 402 | |||
AK170735.1 | - | 4 | 674 | |||
AK171130.1 | - | 3 | 375 | |||
AK171590.1 | - | 3 | 379 | |||
BC052682.1 | - | 3 | 401 | 30072503 |
Gene Symbol: Cd200r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207244.2 | NP_997127.1 | 3 | 338 | ||
GenBank mRNA | AK089262.1 | - | 5 | 633 | ||
AY230200.1 | - | 3 | 338 | |||
BC131946.1 | - | 3 | 328 | 40130223 | ||
BC131972.1 | - | 3 | 328 | 40130249 |
Gene Symbol: F630003A18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177010.3 | NP_795984.1 | 4 | 382 | ||
GenBank mRNA | AK089168.1 | - | 4 | 453 | ||
AK089193.1 | - | 4 | 383 | |||
AK171165.1 | - | 380 | ||||
BC147203.1 | - | 3 | 221 | 9056268 | ||
BC147204.1 | - | 3 | 221 | 9056267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
