siRNA Details
Gene Symbol: Gsdma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007461.1 | NP_001007462.1 | 3, 4 | 480 | ||
GenBank mRNA | AY679090.1 | - | 3, 4 | 480 |
Gene Symbol: Gsdma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021347.4 | NP_067322.1 | 3, 4 | 491 | ||
GenBank mRNA | AB033595.1 | - | 3, 4 | 480 | ||
AK028645.1 | - | 3, 4 | 492 | |||
AK028698.1 | - | 3, 4 | 492 | |||
AK029062.1 | - | 3, 4 | 475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
